Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF280C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017666.4 | NP_060136.1 | 15 | 2143 | ||
XM_006724765.3 | XP_006724828.1 | 14 | 1947 | |||
GenBank mRNA | AK290373.1 | - | 15 | 2094 | ||
AL133204.1 | - | 14 | 1931 | |||
AL834333.1 | - | 15 | 2074 | |||
BC051728.1 | - | 15 | 2094 | 4828435 | ||
DA441065.1 | - | 2 | 167 |