Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143778.1 | NP_001137250.1 | 11, 12 | 1209 | ||
NM_017707.3 | NP_060177.2 | 12, 13 | 1236 | |||
XM_011541755.1 | XP_011540057.1 | 12, 13 | 1176 | |||
XM_017001685.1 | XP_016857174.1 | 12, 13 | 1170 | |||
XM_017001686.1 | XP_016857175.1 | 12, 13 | 1176 | |||
XM_017001687.1 | XP_016857176.1 | 12, 13 | 1143 | |||
XM_017001688.1 | XP_016857177.1 | 12, 13 | 1132 | |||
XM_017001689.1 | XP_016857178.1 | 12, 13 | 1193 | |||
GenBank mRNA | AB056749.1 | - | 12, 13 | 1157 | ||
AK000253.1 | - | 2, 3 | 150 | |||
AK092291.1 | - | 12, 13 | 1158 | |||
AK295100.1 | - | 11, 12 | 1131 | |||
AK299359.1 | - | 12, 13 | 1132 | |||
BC060786.1 | - | 12, 13 | 1184 | 5298355 |