Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143778.1 | NP_001137250.1 | 12, 13 | 1259 | ||
NM_017707.3 | NP_060177.2 | 13, 14 | 1286 | |||
XM_011541755.1 | XP_011540057.1 | 13, 14 | 1226 | |||
XM_017001685.1 | XP_016857174.1 | 13, 14 | 1220 | |||
XM_017001686.1 | XP_016857175.1 | 13, 14 | 1226 | |||
XM_017001687.1 | XP_016857176.1 | 13, 14 | 1193 | |||
XM_017001688.1 | XP_016857177.1 | 13, 14 | 1182 | |||
XM_017001689.1 | XP_016857178.1 | 13, 14 | 1243 | |||
GenBank mRNA | AB056749.1 | - | 13, 14 | 1207 | ||
AK000253.1 | - | 3, 4 | 200 | |||
AK092291.1 | - | 13, 14 | 1208 | |||
AK295100.1 | - | 12, 13 | 1181 | |||
AK299359.1 | - | 13, 14 | 1182 | |||
BC060786.1 | - | 13, 14 | 1234 | 5298355 |