Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMBRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267782.1 | NP_001254711.1 | 13 | 2824 | ||
NM_001267783.1 | NP_001254712.1 | 11 | 2458 | |||
NM_001300731.1 | NP_001287660.1 | 10 | 2635 | |||
NM_017749.3 | NP_060219.2 | 12 | 2543 | |||
XM_005253009.3 | XP_005253066.1 | 11 | 2811 | |||
XM_005253011.3 | XP_005253068.1 | 10 | 2724 | |||
XM_005253014.3 | XP_005253071.1 | 12 | 2541 | |||
XM_006718259.2 | XP_006718322.1 | 11 | 2692 | |||
XM_006718260.2 | XP_006718323.1 | 10 | 2274 | |||
XM_017018007.1 | XP_016873496.1 | 9 | 2544 | |||
XM_017018008.1 | XP_016873497.1 | 11 | 2811 | |||
GenBank mRNA | AB051523.1 | - | 10 | 2587 | ||
AK000301.1 | - | 13 | 2774 | |||
AK023197.1 | - | 5 | 1066 | |||
AK302808.1 | - | 11 | 2873 | |||
AK304530.1 | - | 8 | 1242 | |||
AL137611.1 | - | 3 | 1056 | |||
AL834190.1 | - | 11 | 2337 | |||
BC045609.1 | - | 12 | 2543 | 5296472 | ||
DQ870924.1 | - | 9 | 2366 |