Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127649.2 | NP_001121121.1 | 2 | 702 | ||
NM_001199319.1 | NP_001186248.1 | 3 | 528 | |||
NM_017929.5 | NP_060399.1 | 3 | 528 | |||
GenBank mRNA | AB089678.1 | - | 2 | 418 | ||
AB103104.1 | - | 2 | 320 | |||
AB103105.1 | - | 2 | 320 | |||
AB103106.1 | - | 2 | 319 | |||
AB103107.1 | - | 2 | 319 | |||
AB103108.1 | - | 2 | 319 | |||
AB103109.1 | - | 2 | 319 | |||
AB103110.1 | - | 2 | 320 | |||
AB242827.1 | - | 2 | 319 | |||
AB242828.1 | - | 2 | 319 | |||
AB242829.1 | - | 2 | 319 | |||
AB242830.1 | - | 2 | 319 | |||
AK000065.1 | - | 3 | 528 | |||
AK000702.1 | - | 3 | 525 | |||
BC016280.1 | - | 3 | 446 | 3910351 | ||
BC047320.1 | - | 2 | 688 | 5247852 | ||
CR456362.1 | - | 2 | 337 | |||
KU178709.1 | - | 2 | 319 | |||
KU178710.1 | - | 2 | 319 |