Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001828.5 | NP_001819.2 | 4 | 410 | ||
GenBank mRNA | AV715261.1 | - | 4 | 409 | ||
AV762486.1 | - | 4 | 407 | |||
BC119711.1 | - | 4 | 361 | 40117193 | ||
BC119712.1 | - | 4 | 361 | 40117194 | ||
FJ613334.1 | - | 4 | 353 | |||
FJ613335.1 | - | 4 | 362 | |||
L01664.1 | - | 4 | 366 |