Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000083.2 | NP_000074.2 | 2 | 349 | ||
NR_046453.1 | 2 | 349 | ||||
XM_017011739.1 | XP_016867228.1 | 1 | 207 | |||
XM_017011740.1 | XP_016867229.1 | 1 | 207 | |||
GenBank mRNA | BC112156.1 | - | 2 | 314 | 8327624 | |
BC113495.1 | - | 2 | 314 | 8322547 | ||
BC143796.1 | - | 2 | 314 | |||
Z25884.1 | - | 2 | 349 |