Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018140.3 | NP_060610.2 | 10 | 1652 | ||
XM_005248322.3 | XP_005248379.1 | 10 | 1574 | |||
XM_011514063.1 | XP_011512365.1 | 10 | 1598 | |||
XM_011514064.2 | XP_011512366.1 | 10 | 1705 | |||
XM_017009626.1 | XP_016865115.1 | 9 | 1486 | |||
XM_017009627.1 | XP_016865116.1 | 9 | 1446 | |||
GenBank mRNA | AB040952.1 | - | 10 | 1590 | ||
AK001427.1 | - | 10 | 1595 | |||
BC000132.2 | - | 10 | 1591 | 2899944 | ||
BC001750.1 | - | 10 | 1578 |