Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other COA1 SIRNA Assays ›
Gene Symbol:
COA1
Gene Name:
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases:
C7orf44, MITRAC15
Chromosome Location:
Chr.7: 43608452 - 43729541 on Build GRCh38
UniGene ID:
Hs.268887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31397
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: COA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321197.1
NP_001308126.1
4
476
NM_001321198.1
NP_001308127.1
4
357
NM_001321199.1
NP_001308128.1
4
321
NM_001321200.1
NP_001308129.1
4
377
NM_001321201.1
NP_001308130.1
3
249
NM_001321202.1
NP_001308131.1
4
357
NM_001321203.1
NP_001308132.1
3
256
NM_001321204.1
NP_001308133.1
4
321
NM_001321205.1
NP_001308134.1
4
476
NM_018224.3
NP_060694.2
3
256
NR_047701.1
4
321
NR_135580.1
4
357
NR_135581.1
4
357
NR_135582.1
3
256
NR_135583.1
3
256
XM_005249807.4
XP_005249864.1
3
320
XM_006715752.2
XP_006715815.1
4
288
XM_017012413.1
XP_016867902.1
3
247
XM_017012414.1
XP_016867903.1
3
247
XM_017012415.1
XP_016867904.1
4
349
XM_017012416.1
XP_016867905.1
3
247
XM_017012417.1
XP_016867906.1
3
247
XM_017012418.1
XP_016867907.1
3
247
XM_017012419.1
XP_016867908.1
3
247
XM_017012420.1
XP_016867909.1
4
349
XM_017012421.1
XP_016867910.1
4
471
XM_017012422.1
XP_016867911.1
4
360
XM_017012423.1
XP_016867912.1
4
280
XM_017012424.1
XP_016867913.1
3
263
XM_017012425.1
XP_016867914.1
4
364
XM_017012426.1
XP_016867915.1
4
483
XM_017012427.1
XP_016867916.1
5
438
XM_017012428.1
XP_016867917.1
3
263
XM_017012429.1
XP_016867918.1
2
157
XM_017012430.1
XP_016867919.1
4
349
XM_017012431.1
XP_016867920.1
3
263
XM_017012432.1
XP_016867921.1
3
263
GenBank mRNA
AK001665.1
-
4
290
AK021905.1
-
4
426
AK023007.1
-
4
273
AK023053.1
-
4
289
AK097203.1
-
2
1787
AK293043.1
-
4
299
AK308911.1
-
3
1284
AK309315.1
-
4
299
BC056884.1
-
4
248
6159045
BG028456.1
-
278
BG334368.1
-
4
262
BG548154.1
-
4
313
BM918741.1
-
3
229
BP373150.1
-
5
494
BP377872.1
-
3
257
BQ231567.1
-
3
194
DA099188.1
-
4
296
DA107764.1
-
3
217
DA742258.1
-
3
238
DB003365.1
-
4
433
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
translation
mitochondrial respiratory chain complex I assembly
mitochondrial respiratory chain complex IV assembly
RNA binding
structural constituent of ribosome
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0