Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP5M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018229.3 | NP_060699.3 | 4, 5 | 1518 | ||
NR_026895.1 | 3, 4 | 872 | ||||
XM_006720196.2 | XP_006720259.1 | 4, 5 | 1147 | |||
XM_011536940.2 | XP_011535242.1 | 5, 6 | 1490 | |||
GenBank mRNA | AK001675.1 | - | 4, 5 | 1363 | ||
AK172824.1 | - | 3, 4 | 580 | |||
AK225345.1 | - | 4, 5 | 1365 | |||
AK303999.1 | - | 5, 6 | 1392 | |||
AL136685.1 | - | 4, 5 | 1497 |