Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP5M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018229.3 | NP_060699.3 | 2 | 758 | ||
XM_006720196.2 | XP_006720259.1 | 2 | 387 | |||
XM_011536940.2 | XP_011535242.1 | 3 | 730 | |||
GenBank mRNA | AK001675.1 | - | 2 | 603 | ||
AK225345.1 | - | 2 | 605 | |||
AK303999.1 | - | 3 | 632 | |||
AL136685.1 | - | 2 | 737 | |||
BC013174.1 | - | 2 | 722 | 3924075 |