Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143996.1 | NP_001137468.1 | 9, 10 | 1274 | ||
NM_001143997.1 | NP_001137469.1 | 8, 9 | 1069 | |||
NM_018245.2 | NP_060715.2 | 10, 11 | 1445 | |||
XM_011539946.1 | XP_011538248.1 | 10, 11 | 1330 | |||
XM_011539947.1 | XP_011538249.1 | 10, 11 | 1536 | |||
XM_017016401.1 | XP_016871890.1 | 10, 11 | 1720 | |||
XM_017016402.1 | XP_016871891.1 | 10, 11 | 1347 | |||
XM_017016403.1 | XP_016871892.1 | 8, 9 | 1093 | |||
GenBank mRNA | AB033116.3 | - | 9, 10 | 1308 | ||
AK001713.1 | - | 10, 11 | 1389 | |||
AK290226.1 | - | 9, 10 | 1418 | |||
AK296551.1 | - | 9, 10 | 1183 | |||
AK303729.1 | - | 8, 9 | 1016 | |||
AK307759.1 | - | 9, 10 | 1204 | |||
BC026320.1 | - | 10, 11 | 1364 | 4797814 |