Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286.3 | NP_001277.1 | 3 | 339 | ||
NR_046428.1 | 3 | 339 | ||||
GenBank mRNA | AK289999.1 | - | 3 | 244 | ||
AK307098.1 | - | 3 | 194 | |||
BC117420.1 | - | 3 | 196 | 40125971 | ||
BC117424.1 | - | 3 | 196 | 40125975 | ||
D28475.2 | - | 3 | 224 | |||
DA229615.1 | - | 3 | 339 | |||
DA935587.1 | - | 3 | 285 | |||
X83378.1 | - | 3 | 198 | |||
X96391.1 | - | 3 | 172 | |||
X99473.1 | - | 3 | 172 | |||
X99474.1 | - | 3 | 172 | |||
X99475.1 | - | 3 | 172 |