Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199198.2 | NP_001186127.1 | 11 | 1321 | ||
NM_018309.4 | NP_060779.2 | 11 | 1321 | |||
XM_011512974.2 | XP_011511276.1 | 11 | 1184 | |||
XM_017006841.1 | XP_016862330.1 | 11 | 1152 | |||
GenBank mRNA | AB449910.1 | - | 11 | 1119 | ||
AK001750.1 | - | 8 | 807 | |||
AK001908.1 | - | 11 | 1126 | |||
AL713684.1 | - | 3 | 202 | |||
AY740521.1 | - | 1 | 27 | |||
BC010221.1 | - | 2 | 64 | |||
BC020955.1 | - | 11 | 1115 | 3847561 | ||
BX648032.1 | - | 11 | 1113 |