Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TDP1 SIRNA Assays ›
Gene Symbol:
TDP1
Gene Name:
tyrosyl-DNA phosphodiesterase 1
Gene Aliases:
-
Chromosome Location:
Chr.14: 89954935 - 90044764 on Build GRCh38
UniGene ID:
Hs.209945
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31480
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TDP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008744.1
NP_001008744.1
11
1427
NM_018319.3
NP_060789.2
12
1650
XM_005267847.2
XP_005267904.1
12
2324
XM_005267848.2
XP_005267905.1
11
1668
XM_006720197.3
XP_006720260.1
12
1891
XM_006720198.3
XP_006720261.1
11
1546
XM_006720199.1
XP_006720262.1
11
1032
XM_011536942.2
XP_011535244.1
11
2067
XM_011536943.2
XP_011535245.1
11
1460
XM_011536944.1
XP_011535246.1
12
1651
XM_017021439.1
XP_016876928.1
12
1558
XM_017021440.1
XP_016876929.1
11
1450
XM_017021441.1
XP_016876930.1
11
1639
XM_017021442.1
XP_016876931.1
9
1276
XM_017021443.1
XP_016876932.1
9
775
XM_017021444.1
XP_016876933.1
9
776
XM_017021445.1
XP_016876934.1
9
1276
XM_017021446.1
XP_016876935.1
10
1082
GenBank mRNA
AF182003.1
-
2
96
AK001952.1
-
11
1391
AK023514.1
-
1
13
AK093235.1
-
13
2458
AK315549.1
-
11
1375
AL832288.1
-
3
369
BC006083.1
-
14
1814
BC015474.1
-
12
1601
3900062
BC107877.1
-
12
1654
6597752
BX161451.1
-
11
1562
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
single strand break repair
DNA repair
double-strand break repair
nucleic acid phosphodiester bond hydrolysis
double-stranded DNA binding
single-stranded DNA binding
exonuclease activity
protein binding
3'-tyrosyl-DNA phosphodiesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0