Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM63A SIRNA Assays ›
Gene Symbol:
FAM63A
Gene Name:
family with sequence similarity 63 member A
Gene Aliases:
-
Chromosome Location:
Chr.1: 150996737 - 151008378 on Build GRCh38
UniGene ID:
Hs.743952
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31524
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM63A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040217.2
NP_001035307.1
7, 8
1284
NM_001163258.1
NP_001156730.1
8, 9
1347
NM_001163259.1
NP_001156731.1
7, 8
1249
NM_001163260.1
NP_001156732.1
7, 8
1160
NM_001319998.1
NP_001306927.1
8, 9
2664
NM_018379.4
NP_060849.2
8, 9
1948
XM_005245321.3
XP_005245378.1
7, 8
1925
XM_005245325.3
XP_005245382.1
7, 8
1280
XM_005245327.3
XP_005245384.1
6, 7
1121
XM_005245328.3
XP_005245385.1
6, 7
1118
XM_011509757.2
XP_011508059.1
7, 8
1925
XM_011509759.2
XP_011508061.1
6, 7
1501
XM_017001770.1
XP_016857259.1
7, 8
1922
XM_017001771.1
XP_016857260.1
7, 8
1660
XM_017001772.1
XP_016857261.1
7, 8
1660
XM_017001773.1
XP_016857262.1
7, 8
1922
XM_017001774.1
XP_016857263.1
7, 8
1660
XM_017001775.1
XP_016857264.1
6, 7
1763
XM_017001776.1
XP_016857265.1
6, 7
1763
XM_017001777.1
XP_016857266.1
7, 8
1274
GenBank mRNA
AB037811.1
-
6, 7
1133
AK002142.1
-
7, 8
1010
AK125493.1
-
7, 8
984
AK125959.1
-
8, 9
2664
AK301946.1
-
8, 9
1347
AK303962.1
-
7, 8
937
BC032321.1
-
7, 8
1424
5240620
BC075812.1
-
8, 9
1935
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0