Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018396.2 | NP_060866.2 | 9 | 1812 | ||
GenBank mRNA | BC064929.1 | - | 9 | 1773 | ||
BC107586.1 | - | 8 | 1697 | 6494734 |