Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040151.1 | NP_001035241.1 | 5 | 934 | ||
NM_018400.3 | NP_060870.1 | 4 | 1333 | |||
XM_011542897.1 | XP_011541199.1 | 5 | 934 | |||
GenBank mRNA | AB032984.1 | - | 5 | 574 | ||
AJ243396.2 | - | 4 | 902 | |||
AK314513.1 | - | 5 | 921 | |||
AL136589.1 | - | 4 | 1330 | |||
BC117282.1 | - | 4 | 620 | 40125833 | ||
BC126265.1 | - | 4 | 620 | 8991981 |