Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040428.3 | NP_001035518.1 | 8 | 1151 | ||
NM_018418.4 | NP_060888.2 | 9 | 1247 | |||
XM_005267851.1 | XP_005267908.1 | 9 | 1221 | |||
XM_005267852.1 | XP_005267909.1 | 8 | 1150 | |||
XM_005267854.1 | XP_005267911.1 | 10 | 1283 | |||
XM_005267855.1 | XP_005267912.1 | 9 | 1216 | |||
XM_006720204.1 | XP_006720267.1 | 9 | 1221 | |||
XM_006720205.1 | XP_006720268.1 | 9 | 1221 | |||
XM_011536951.1 | XP_011535253.1 | 9 | 1415 | |||
XM_011536952.1 | XP_011535254.1 | 7 | 1033 | |||
XM_011536953.1 | XP_011535255.1 | 8 | 1354 | |||
XM_017021452.1 | XP_016876941.1 | 9 | 1412 | |||
XM_017021453.1 | XP_016876942.1 | 9 | 1381 | |||
XM_017021454.1 | XP_016876943.1 | 10 | 1280 | |||
XM_017021455.1 | XP_016876944.1 | 9 | 1213 | |||
XM_017021456.1 | XP_016876945.1 | 10 | 1226 | |||
XM_017021457.1 | XP_016876946.1 | 8 | 1351 | |||
GenBank mRNA | AF144487.3 | - | 9 | 1227 | ||
AF144488.2 | - | 7 | 955 | |||
AK055864.1 | - | 8 | 2819 | |||
AK225746.1 | - | 10 | 1221 | |||
AK290631.1 | - | 8 | 1122 | |||
AL136604.1 | - | 8 | 1131 | |||
BC008656.1 | - | 4 | 679 | |||
BC090875.1 | - | 9 | 1534 | 30383306 | ||
CR533563.1 | - | 8 | 976 | |||
CR936777.1 | - | 9 | 1193 | |||
EU668353.1 | - | 9 | 1223 |