Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018447.3 | NP_060917.1 | 4, 5 | 785 | ||
XM_005265321.3 | XP_005265378.1 | 4, 5 | 653 | |||
GenBank mRNA | AF157321.1 | - | 3, 4 | 497 | ||
AK172843.1 | - | 3, 4 | 493 | |||
AK222953.1 | - | 3, 4 | 495 | |||
AK312008.1 | - | 3, 4 | 492 | |||
BC022807.1 | - | 3, 4 | 480 | 4897577 |