Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018453.3 | NP_060923.2 | 4 | 515 | ||
XM_011536958.1 | XP_011535260.1 | 4 | 793 | |||
XM_017021460.1 | XP_016876949.1 | 4 | 515 | |||
GenBank mRNA | AF217512.1 | - | 4 | 501 | ||
AK000585.1 | - | 4 | 479 | |||
AK225467.1 | - | 4 | 470 | |||
AK225477.1 | - | 4 | 479 | |||
AK225991.1 | - | 4 | 479 | |||
AY157300.1 | - | 4 | 497 | |||
AY157301.1 | - | 4 | 497 | |||
AY869694.1 | - | 4 | 438 | |||
BC001245.1 | - | 4 | 488 | 3460193 | ||
CD702290.1 | - | 4 | 544 |