Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307985.1 | NP_001294914.1 | 3 | 1033 | ||
NM_018484.3 | NP_060954.1 | 3 | 1033 | |||
XM_011545167.1 | XP_011543469.1 | 2 | 683 | |||
GenBank mRNA | AB026116.1 | - | 3 | 686 | ||
AK075127.1 | - | 1 | 59 | |||
AK075224.1 | - | 3 | 379 | |||
AK129930.1 | - | 3 | 945 | |||
AK222869.1 | - | 3 | 691 | |||
AK290791.1 | - | 3 | 1016 | |||
AK296083.1 | - | 2 | 616 | |||
AK309931.1 | - | 3 | 693 | |||
BC034384.1 | - | 3 | 1008 | 5190509 |