Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018652.4 | NP_061122.4 | 10 | 835 | ||
XM_017022406.1 | XP_016877895.1 | 15 | 1723 |
Gene Symbol: GOLGA6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038640.2 | NP_001033729.2 | 10 | 835 | ||
GenBank mRNA | AF263742.1 | - | 10 | 835 | ||
AK292574.1 | - | 10 | 816 | |||
BC131515.1 | - | 10 | 815 |
Gene Symbol: GOLGA6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164404.1 | NP_001157876.1 | 10 | 835 | ||
XM_017022519.1 | XP_016878008.1 | 10 | 989 |
Gene Symbol: GOLGA6D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145224.1 | NP_001138696.1 | 10 | 829 |
Gene Symbol: LOC645752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027024.1 | 10 | 820 |