Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018682.3 | NP_061152.3 | 26 | 5672 | ||
NM_182931.2 | NP_891847.1 | 27 | 5820 | |||
XM_005250493.1 | XP_005250550.1 | 26 | 5739 | |||
XM_011516400.1 | XP_011514702.1 | 28 | 5946 | |||
XM_017012435.1 | XP_016867924.1 | 27 | 5820 | |||
GenBank mRNA | AF519459.1 | - | 26 | 5487 | ||
AY113691.1 | - | 1 | 92 | |||
AY147037.1 | - | 27 | 5820 | |||
AY157990.1 | - | 24 | 5046 | |||
AY222296.1 | - | 2 | 393 | |||
AY438698.1 | - | 2 | 307 |