Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018686.5 | NP_061156.1 | 4 | 708 | ||
NR_135117.1 | 4 | 708 | ||||
GenBank mRNA | AF271388.1 | - | 4 | 596 | ||
AF397212.1 | - | 4 | 578 | |||
AK022927.1 | - | 4 | 618 | |||
AK222981.1 | - | 4 | 607 | |||
AL832975.1 | - | 4 | 656 | |||
BC016609.1 | - | 4 | 657 | 3448277 |