Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018688.4 | NP_061158.1 | 6 | 420 | ||
XM_005273573.3 | XP_005273630.1 | 5 | 326 | |||
XM_011544586.2 | XP_011542888.1 | 7 | 505 | |||
XM_011544587.1 | XP_011542889.1 | 6 | 420 | |||
GenBank mRNA | AF271732.1 | - | 6 | 383 | ||
AK001289.1 | - | 4 | 282 | |||
AK023980.1 | - | 6 | 367 | |||
AK090446.1 | - | 3 | 929 | |||
AK222470.1 | - | 6 | 392 | |||
BC001223.1 | - | 5 | 304 | 3534031 | ||
BC009824.2 | - | 6 | 354 | 3953604 |