Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242617.1 | NP_001229546.1 | 1, 2 | 196 | ||
NM_018698.4 | NP_061168.2 | 2, 3 | 286 | |||
GenBank mRNA | AF201942.1 | - | 1, 2 | 163 | ||
AF246127.1 | - | 19 | ||||
AJ277591.1 | - | 19 | ||||
AK023289.1 | - | 2, 3 | 286 | |||
BC014888.1 | - | 1, 2 | 150 | |||
BC120984.1 | - | 2, 3 | 275 | 40116082 | ||
BC120985.1 | - | 2, 3 | 275 | 40116088 | ||
DA442845.1 | - | 1, 2 | 198 |