Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034023.1 | NP_001029195.1 | 6 | 762 | ||
NM_001034024.1 | NP_001029196.1 | 6 | 722 | |||
NM_019100.4 | NP_061973.1 | 5 | 943 | |||
XM_005271037.4 | XP_005271094.1 | 5 | 933 | |||
XM_005271039.1 | XP_005271096.1 | 6 | 833 | |||
XM_005271040.1 | XP_005271097.1 | 6 | 710 | |||
XM_006710771.3 | XP_006710834.1 | 4 | 612 | |||
XM_011541784.2 | XP_011540086.1 | 6 | 767 | |||
XM_011541785.2 | XP_011540087.1 | 6 | 787 | |||
XM_017001807.1 | XP_016857296.1 | 6 | 818 | |||
XM_017001808.1 | XP_016857297.1 | 6 | 821 | |||
GenBank mRNA | AB037846.1 | - | 6 | 727 | ||
AF265228.1 | - | 5 | 642 | |||
AK021605.1 | - | 6 | 706 | |||
AK054954.1 | - | 2 | 1560 | |||
AK092898.1 | - | 2 | 3727 | |||
AK096252.1 | - | 2 | 2330 | |||
AK289366.1 | - | 6 | 745 | |||
AK293598.1 | - | 6 | 869 | |||
AK298793.1 | - | 6 | 705 | |||
AL136657.1 | - | 6 | 724 | |||
AL137200.1 | - | 6 | 726 | |||
BC002855.2 | - | 6 | 711 | 3635724 | ||
BC008053.2 | - | 5 | 905 | 3834056 | ||
BX537895.1 | - | 2 | 2881 |