Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008405.2 | NP_001008405.1 | 6 | 659 | ||
NM_018844.3 | NP_061332.2 | 7 | 984 | |||
NR_027830.1 | 6 | 878 | ||||
XM_006716051.2 | XP_006716114.1 | 7 | 980 | |||
XM_011516406.2 | XP_011514708.1 | 7 | 743 | |||
XM_011516407.1 | XP_011514709.1 | 7 | 980 | |||
XM_011516408.2 | XP_011514710.1 | 7 | 980 | |||
XM_011516409.2 | XP_011514711.1 | 7 | 980 | |||
XM_017012436.1 | XP_016867925.1 | 7 | 980 | |||
XM_017012437.1 | XP_016867926.1 | 7 | 980 | |||
XM_017012438.1 | XP_016867927.1 | 7 | 980 | |||
GenBank mRNA | AA826200.1 | - | 3 | 280 | ||
AF126020.1 | - | 7 | 694 | |||
AK289401.1 | - | 7 | 741 | |||
AK294824.1 | - | 6 | 609 | |||
AK298594.1 | - | 4 | 385 | |||
AW978976.1 | - | 3 | 304 | |||
BC008478.1 | - | 7 | 696 | 4294273 | ||
BG818947.1 | - | 2 | 175 | |||
BM023571.1 | - | 4 | 454 | |||
BP261060.1 | - | 4 | 323 | |||
BQ230678.1 | - | 261 | ||||
BT006981.1 | - | 6 | 645 | |||
BX647653.1 | - | 7 | 694 | |||
CK820004.1 | - | 5 | 516 |