Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282175.1 | NP_001269104.1 | 5 | 517 | ||
NM_001282176.1 | NP_001269105.1 | 5 | 541 | |||
NM_001294.3 | NP_001285.1 | 5 | 668 | |||
GenBank mRNA | AF037339.1 | - | 5 | 511 | ||
AK074935.1 | - | 5 | 512 | |||
AK223553.1 | - | 5 | 512 | |||
AK293314.1 | - | 5 | 533 | |||
AK304480.1 | - | 5 | 500 | |||
AK310826.1 | - | 5 | 541 | |||
AK316094.1 | - | 5 | 537 | |||
BC004865.2 | - | 5 | 547 | 3840991 | ||
BC012359.1 | - | 5 | 501 | 4685026 | ||
BI916245.1 | - | 5 | 514 | |||
BT007262.1 | - | 5 | 496 | |||
DC313765.1 | - | 5 | 517 | |||
DC383798.1 | - | 5 | 541 | |||
KU177986.1 | - | 5 | 496 | |||
KU177987.1 | - | 5 | 496 |