Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018907.3 | NP_061730.1 | 1 | 895 | ||
NM_031500.2 | NP_113688.1 | 1 | 895 | |||
GenBank mRNA | AB208925.1 | - | 1 | 895 | ||
AF152312.1 | - | 1 | 781 | |||
AF152482.1 | - | 1 | 781 | |||
BC112102.1 | - | 1 | 867 | 8327570 | ||
BC113609.1 | - | 1 | 867 | 8322661 |