Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C21orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162495.2 | NP_001155967.2 | 4 | 664 | ||
NM_001162496.2 | NP_001155968.2 | 2 | 508 | |||
NM_019596.5 | NP_062542.5 | 3 | 589 | |||
GenBank mRNA | AF231922.1 | - | 3 | 495 | ||
AK290983.1 | - | 3 | 532 |