Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127244.1 | NP_001120716.1 | 3 | 616 | ||
NM_001127245.1 | NP_001120717.1 | 2 | 240 | |||
NM_019594.3 | NP_062540.2 | 3 | 347 | |||
XM_005252095.2 | XP_005252152.1 | 3 | 477 | |||
XM_005252096.3 | XP_005252153.1 | 3 | 1640 | |||
XM_006717186.2 | XP_006717249.1 | 4 | 692 | |||
XM_011518863.1 | XP_011517165.1 | 3 | 283 | |||
XM_011518864.2 | XP_011517166.1 | 4 | 401 | |||
XM_011518865.2 | XP_011517167.1 | 2 | 119 | |||
GenBank mRNA | AB037858.1 | - | 2 | 156 | ||
AK024649.1 | - | 2 | 177 | |||
AK289842.1 | - | 3 | 616 | |||
AY143166.1 | - | 1 | 93 | |||
BC051322.1 | - | 3 | 259 | 6250713 | ||
BX376144.2 | - | 3 | 291 | |||
DA243590.1 | - | 3 | 347 |