Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019644.3 | NP_062618.2 | 1 | 319 | ||
GenBank mRNA | AK302407.1 | - | 1 | 273 | ||
BC032799.1 | - | 1 | 80 | 5170634 | ||
D78334.1 | - | 1 | 74 | |||
DB442002.1 | - | 1 | 270 | |||
DB447815.1 | - | 1 | 314 |