Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286130.1 | NP_001273059.1 | 21, 22 | 2214 | ||
NM_001297.4 | NP_001288.3 | 21, 22 | 2232 | |||
XM_011522870.2 | XP_011521172.1 | 9, 10 | 1501 | |||
GenBank mRNA | AF042498.1 | - | 21, 22 | 2219 | ||
U58837.1 | - | 21, 22 | 2224 |