Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077238.1 | NP_001070706.1 | 4 | 478 | ||
NM_152988.2 | NP_694533.1 | 4 | 478 | |||
XM_011528138.2 | XP_011526440.1 | 4 | 537 | |||
XM_011528139.1 | XP_011526441.1 | 3 | 332 | |||
XM_017027016.1 | XP_016882505.1 | 4 | 537 | |||
XM_017027017.1 | XP_016882506.1 | 5 | 668 | |||
XM_017027018.1 | XP_016882507.1 | 3 | 332 | |||
XM_017027019.1 | XP_016882508.1 | 5 | 668 | |||
GenBank mRNA | AB040965.1 | - | 4 | 427 | ||
AJ345027.1 | - | 4 | 431 | |||
AJ420897.1 | - | 4 | 427 | |||
AK226130.1 | - | 1 | 21 | |||
AK308973.1 | - | 5 | 547 | |||
AK309122.1 | - | 4 | 552 | |||
AY169315.1 | - | 4 | 426 | |||
BC001788.1 | - | 4 | 412 | 3352341 | ||
BC028391.2 | - | 4 | 415 | 4822524 | ||
BC093046.1 | - | 4 | 478 | 30395602 |