Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082533.1 | NP_001076002.1 | 8 | 1793 | ||
NM_001082534.1 | NP_001076003.1 | 8 | 1358 | |||
NM_020178.4 | NP_064563.1 | 7 | 1667 | |||
XM_017024878.1 | XP_016880367.1 | 5 | 521 | |||
GenBank mRNA | AB001597.1 | - | 7 | 681 | ||
AB036836.1 | - | 8 | 1583 | |||
AF288385.1 | - | 8 | 1349 | |||
AK294652.1 | - | 6 | 1038 | |||
AK313001.1 | - | 7 | 1243 | |||
AL161812.1 | - | 7 | 683 | |||
AY358509.1 | - | 7 | 961 | |||
BC020577.1 | - | 8 | 1325 | 4181377 | ||
BC029865.1 | - | 8 | 1327 | |||
BC047456.1 | - | 7 | 1580 | 5276935 | ||
BC068462.1 | - | 7 | 1626 | 5260321 | ||
CR749607.1 | - | 8 | 1129 |