Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082533.1 | NP_001076002.1 | 6 | 1591 | ||
NM_001082534.1 | NP_001076003.1 | 6 | 1156 | |||
NM_020178.4 | NP_064563.1 | 5 | 1465 | |||
XM_017024878.1 | XP_016880367.1 | 3 | 319 | |||
GenBank mRNA | AB001597.1 | - | 5 | 479 | ||
AB036836.1 | - | 6 | 1381 | |||
AF288385.1 | - | 6 | 1147 | |||
AK294652.1 | - | 4 | 836 | |||
AK313001.1 | - | 5 | 1041 | |||
AL161812.1 | - | 5 | 481 | |||
AY358509.1 | - | 5 | 759 | |||
BC020577.1 | - | 6 | 1123 | 4181377 | ||
BC029865.1 | - | 6 | 1125 | |||
BC047456.1 | - | 5 | 1378 | 5276935 | ||
BC068462.1 | - | 5 | 1424 | 5260321 | ||
CR749607.1 | - | 6 | 927 |