Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020188.3 | NP_064573.1 | 3 | 328 | ||
XM_017023461.1 | XP_016878950.1 | 4 | 516 | |||
XM_017023462.1 | XP_016878951.1 | 4 | 531 | |||
XM_017023463.1 | XP_016878952.1 | 3 | 456 | |||
XM_017023467.1 | XP_016878956.1 | 5 | 411 | |||
XM_017023468.1 | XP_016878957.1 | 4 | 321 | |||
XM_017023469.1 | XP_016878958.1 | 4 | 395 | |||
XM_017023470.1 | XP_016878959.1 | 4 | 319 | |||
GenBank mRNA | AF201935.1 | - | 3 | 304 | ||
AK123193.1 | - | 2 | 1683 | |||
BC032631.1 | - | 3 | 332 | 5399193 |