Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193557.1 | NP_001180486.1 | 3, 4 | 231 | ||
NM_020189.5 | NP_064574.1 | 3, 4 | 303 | |||
GenBank mRNA | AF173296.1 | - | 3, 4 | 373 | ||
AF201940.1 | - | 3, 4 | 318 | |||
AK000487.1 | - | 3, 4 | 207 | |||
AK290752.1 | - | 3, 4 | 247 | |||
AK316560.1 | - | 3, 4 | 298 | |||
BC007870.2 | - | 3, 4 | 219 | 4303290 | ||
BX648621.1 | - | 1, 2 | 1052 | |||
CR457183.1 | - | 2, 3 | 157 | |||
DA548665.1 | - | 3, 4 | 177 | |||
DB168034.1 | - | 3, 4 | 303 |