Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MFF SIRNA Assays ›
Gene Symbol:
MFF
LOC644762
Gene Name:
mitochondrial fission factor
mitochondrial fission factor pseudogene
Gene Aliases:
C2orf33, GL004
-
Chromosome Location:
Chr.2: 227325151 - 227357836 on Build GRCh38
Chromosome Location:
Chr.5: 149931358 - 149932977 on Build GRCh38
UniGene ID:
Hs.471528
Hs.730366
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s32460
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MFF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001277061.1
NP_001263990.1
10
1415
NM_001277062.1
NP_001263991.1
8
1162
NM_001277063.1
NP_001263992.1
7
1018
NM_001277064.1
NP_001263993.1
7
1003
NM_001277065.1
NP_001263994.1
6
943
NM_001277066.1
NP_001263995.1
5
831
NM_001277067.1
NP_001263996.1
7
848
NM_001277068.1
NP_001263997.1
7
768
NM_020194.5
NP_064579.3
9
1152
NR_102266.1
4
610
XM_005246681.3
XP_005246738.1
9
1275
XM_005246682.1
XP_005246739.1
9
1260
XM_005246683.1
XP_005246740.1
9
1157
XM_005246684.1
XP_005246741.1
8
1200
XM_005246685.3
XP_005246742.1
9
1176
XM_005246686.3
XP_005246743.1
8
1116
XM_005246687.3
XP_005246744.1
8
1101
XM_005246688.1
XP_005246745.1
7
1041
XM_006712637.1
XP_006712700.1
8
1059
XM_006712638.1
XP_006712701.1
7
984
XM_006712639.2
XP_006712702.1
6
825
XM_011511500.1
XP_011509802.1
10
1207
XM_011511501.1
XP_011509803.1
9
1228
XM_017004512.1
XP_016860001.1
9
1324
XM_017004513.1
XP_016860002.1
7
1022
XM_017004514.1
XP_016860003.1
6
924
XM_017004515.1
XP_016860004.1
6
840
GenBank mRNA
AF226049.1
-
5
586
AF246239.1
-
9
1152
AF258660.1
-
10
1270
AK026137.1
-
8
1063
AL517791.3
-
7
879
AL833032.1
-
3
792
BC000797.1
-
6
675
3451868
BC093024.1
-
7
901
30344432
BQ653061.1
-
5
573
DA737008.1
-
7
742
W58482.1
-
4
532
Gene Symbol: LOC644762
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_110534.1
1
826
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial fission
release of cytochrome c from mitochondria
protein targeting to mitochondrion
mitochondrial fusion
regulation of mitochondrion organization
peroxisome fission
mitochondrial fragmentation involved in apoptotic process
protein homooligomerization
mitochondrion morphogenesis
positive regulation of mitochondrial fission
positive regulation of release of cytochrome c from mitochondria
positive regulation of protein targeting to membrane
regulation of peroxisome organization
protein binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0