Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020232.4 | NP_064617.2 | 6 | 778 | ||
NM_147163.1 | NP_671692.1 | 6 | 636 | |||
GenBank mRNA | AF068295.1 | - | 6 | 636 | ||
AF161378.1 | - | 6 | 632 | |||
AF168712.1 | - | 6 | 710 | |||
AF276707.1 | - | 6 | 1338 | |||
AK025410.1 | - | 2 | 124 | |||
AK057005.1 | - | 6 | 710 | |||
AK225882.1 | - | 6 | 745 | |||
BC013356.2 | - | 6 | 701 | 3640209 | ||
CR457181.1 | - | 5 | 538 |