Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267812.1 | NP_001254741.1 | 2 | 320 | ||
NM_001267814.1 | NP_001254743.1 | 2 | 324 | |||
NM_020246.3 | NP_064631.2 | 2 | 320 | |||
XM_005250502.2 | XP_005250559.1 | 2 | 323 | |||
XM_006716054.2 | XP_006716117.1 | 2 | 305 | |||
XM_006716055.2 | XP_006716118.1 | 1 | 202 | |||
XM_011516414.1 | XP_011514716.1 | 2 | 302 | |||
GenBank mRNA | AB033284.1 | - | 2 | 247 | ||
AF284422.1 | - | 2 | 287 | |||
AK128873.1 | - | 2 | 270 | |||
AK301411.1 | - | 2 | 320 | |||
BC000154.2 | - | 2 | 306 | 2900382 | ||
DA816969.1 | - | 2 | 285 | |||
DA982886.1 | - | 2 | 320 |