Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020247.4 | NP_064632.2 | 14 | 1796 | ||
XM_005273201.1 | XP_005273258.1 | 14 | 1692 | |||
XM_011544238.1 | XP_011542540.1 | 15 | 1888 | |||
XM_011544239.2 | XP_011542541.1 | 16 | 1911 | |||
XM_011544240.2 | XP_011542542.1 | 14 | 1705 | |||
XM_011544241.2 | XP_011542543.1 | 14 | 1667 | |||
XM_017001852.1 | XP_016857341.1 | 14 | 1886 | |||
GenBank mRNA | AB073905.1 | - | 14 | 1719 | ||
AF218003.1 | - | 6 | 811 | |||
AJ278126.1 | - | 10 | 1014 | |||
AK074693.1 | - | 14 | 1739 | |||
AK090494.1 | - | 14 | 1533 | |||
AK092784.1 | - | 14 | 1519 | |||
AK293571.1 | - | 11 | 1159 | |||
AK297735.1 | - | 11 | 1075 | |||
AK300537.1 | - | 10 | 885 | |||
AK301012.1 | - | 16 | 1717 | |||
BC005171.2 | - | 14 | 1745 | 3604948 | ||
BX648860.1 | - | 14 | 1617 |