Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020247.4 | NP_064632.2 | 12 | 1621 | ||
XM_005273201.1 | XP_005273258.1 | 12 | 1517 | |||
XM_011544238.1 | XP_011542540.1 | 13 | 1713 | |||
XM_011544239.2 | XP_011542541.1 | 14 | 1736 | |||
XM_011544240.2 | XP_011542542.1 | 12 | 1530 | |||
XM_011544241.2 | XP_011542543.1 | 12 | 1492 | |||
XM_017001852.1 | XP_016857341.1 | 12 | 1711 | |||
GenBank mRNA | AF218003.1 | - | 4 | 636 | ||
AJ278126.1 | - | 8 | 839 | |||
AK074693.1 | - | 12 | 1564 | |||
AK090494.1 | - | 12 | 1358 | |||
AK092784.1 | - | 12 | 1344 | |||
AK300537.1 | - | 8 | 710 | |||
BC005171.2 | - | 12 | 1570 | 3604948 |