Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020312.3 | NP_064708.1 | 2 | 223 | ||
GenBank mRNA | AF161444.1 | - | 2 | 154 | ||
AK075438.1 | - | 2 | 224 | |||
AK290627.1 | - | 2 | 153 | |||
AK293585.1 | - | 2 | 157 | |||
AK295793.1 | - | 2 | 166 | |||
AK303527.1 | - | 2 | 146 | |||
BC029341.1 | - | 2 | 157 | |||
BC054340.1 | - | 2 | 157 | 6514554 | ||
BC064946.1 | - | 2 | 158 | 6023461 | ||
BM712215.1 | - | 2 | 211 |