Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308185.1 | NP_001295114.1 | 5 | 1305 | ||
NM_020307.3 | NP_064703.1 | 5 | 1305 | |||
XM_005247648.3 | XP_005247705.1 | 6 | 762 | |||
XM_006713710.3 | XP_006713773.1 | 5 | 749 | |||
XM_011513011.2 | XP_011511313.1 | 6 | 1314 | |||
XM_017006891.1 | XP_016862380.1 | 6 | 868 | |||
XM_017006892.1 | XP_016862381.1 | 6 | 885 | |||
XM_017006893.1 | XP_016862382.1 | 6 | 793 | |||
XM_017006894.1 | XP_016862383.1 | 6 | 885 | |||
XM_017006895.1 | XP_016862384.1 | 6 | 762 | |||
XM_017006896.1 | XP_016862385.1 | 5 | 689 | |||
XM_017006897.1 | XP_016862386.1 | 6 | 837 | |||
GenBank mRNA | AF180920.1 | - | 5 | 693 | ||
AF208843.1 | - | 1 | 126 | |||
AF367476.1 | - | 5 | 639 | |||
AK022974.1 | - | 5 | 692 | |||
AK098021.1 | - | 1 | 2455 | |||
AK122738.1 | - | 5 | 1305 | |||
AY034790.1 | - | 6 | 1052 | |||
BC007081.1 | - | 5 | 692 | 4066376 | ||
BC038394.1 | - | 6 | 753 | |||
BC067812.1 | - | 5 | 681 | 5298716 | ||
BX641146.1 | - | 1 | 1617 | |||
BX647554.1 | - | 1 | 290 |