Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160226.1 | NP_001153698.1 | 3 | 1014 | ||
NM_001160258.1 | NP_001153730.1 | 4 | 1052 | |||
NM_001160259.1 | NP_001153731.1 | 2 | 927 | |||
NM_016083.4 | NP_057167.2 | 2 | 871 | |||
NM_033181.3 | NP_149421.2 | 2 | 526 | |||
XM_006715330.3 | XP_006715393.1 | 2 | 818 | |||
XM_011535424.1 | XP_011533726.1 | 3 | 1358 | |||
XM_011535425.1 | XP_011533727.1 | 4 | 1396 | |||
XM_011535426.1 | XP_011533728.1 | 5 | 1554 | |||
XM_011535428.1 | XP_011533730.1 | 3 | 1205 | |||
XM_017010238.1 | XP_016865727.1 | 4 | 4182 | |||
XM_017010239.1 | XP_016865728.1 | 3 | 2505 | |||
XM_017010240.1 | XP_016865729.1 | 2 | 865 | |||
GenBank mRNA | AF107262.1 | - | 1 | 571 | ||
AK313908.1 | - | 2 | 692 | |||
AY766182.1 | - | 2 | 463 | |||
AY995204.1 | - | 1 | 562 | |||
BC074811.2 | - | 1 | 600 | 30915316 | ||
BC074812.2 | - | 1 | 600 | 30915532 | ||
BC095513.1 | - | 1 | 559 | |||
BC100968.1 | - | 1 | 562 | 40004970 | ||
BC100969.1 | - | 1 | 562 | 40004975 | ||
BC100970.1 | - | 1 | 562 | 40004978 | ||
BC100971.1 | - | 1 | 562 | 40004979 | ||
DQ067455.1 | - | 1 | 562 | |||
KC292264.1 | - | 2 | 526 | |||
X54937.1 | - | 2 | 710 | |||
X81120.1 | - | 1 | 653 | |||
X81121.1 | - | 2 | 395 |
Set Membership: |
G-protein coupled receptor |