Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040194.1 | NP_001035284.1 | 5 | 1136 | ||
NM_001040195.1 | NP_001035285.1 | 4 | 1101 | |||
NM_001040196.1 | NP_001035286.1 | 6 | 1233 | |||
NM_001040197.1 | NP_001035287.1 | 6 | 1254 | |||
NM_020350.4 | NP_065083.3 | 5 | 1157 | |||
XM_011541799.2 | XP_011540101.2 | 6 | 1150 | |||
XM_011541800.2 | XP_011540102.1 | 6 | 1171 | |||
XM_011541802.1 | XP_011540104.1 | 7 | 1247 | |||
GenBank mRNA | AF312374.1 | - | 5 | 1088 | ||
AK290042.1 | - | 5 | 1100 | |||
BC011946.1 | - | 5 | 1042 | |||
BC017328.2 | - | 5 | 1074 | 4546585 | ||
BQ672962.1 | - | 1 | 641 |