Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRASLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020386.4 | NP_065119.2 | 3 | 584 | ||
XM_011513034.1 | XP_011511336.1 | 3 | 584 | |||
XM_011513035.2 | XP_011511337.1 | 3 | 584 | |||
XM_017006923.1 | XP_016862412.1 | 4 | 465 | |||
GenBank mRNA | AB030816.1 | - | 3 | 389 | ||
AB510981.1 | - | 2 | 178 | |||
BC048095.1 | - | 3 | 587 | 4830228 |